missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 197320050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bis(triphenylphosphine)palladium(II) chloride | |
260°C | |
Authentic | |
C36H30Cl2P2Pd | |
MFCD00009593 | |
06,59; 07,95; 08,151; 17,111 | |
Solubility in water: insoluble. | |
[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
701.90 | |
701.89 | |
Analysis |
13965-03-2 | |
Yellow | |
Glass bottle | |
[(C6H5)3P]2PdCl2 | |
5 g | |
bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g | |
YNHIGQDRGKUECZ-UHFFFAOYSA-L | |
ethane;methane;palladium(2+);triphenylphosphane;dichloride | |
131664180 | |
15% Pd | |
Powder |
Chemical Identifiers
13965-03-2 | |
701.90 | |
YNHIGQDRGKUECZ-UHFFFAOYSA-L | |
131664180 | |
[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C36H30Cl2P2Pd | |
MFCD00009593 | |
bis triphenylphosphine palladium ii dichloride, bis triphenylphosphine palladium ii chloride, dichlorobis triphenylphosphine palladium, bis triphenylphosphine palladium chloride, bis triphenylphosphine palladiuml ii dichloride, palladium 2+ ; triphenylphosphane; dichloride, trans-dichlorobis triphenylphosphine palladium ii 1g | |
ethane;methane;palladium(2+);triphenylphosphane;dichloride |
Safety and Handling
GHS H Statement
May cause long lasting harmful effects to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
Warning
EINECSNumber : 237-744-2
RUO – Research Use Only