Please login to your online account to display your discounted pricing

Bis(triphenylphosphine)palladium(II) chloride, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  299250050

 View more versions of this product

Catalog No. AC299250050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay 14.8 to 15.5% (Pd)
Formula Weight 701.89
Physical Form Liquid
Quantity 5g
Color Clear colorless
Fieser 06,59; 07,95; 08,151; 17,111
Packaging Glass bottle
Chemical Name or Material Bis(triphenylphosphine)palladium(II) chloride
CAS 13965-03-2
Assay Percent Range 98%
Synonym Dichlorobis(triphenylphosphine)palladium(II)
Molecular Formula C36H30Cl2P2Pd
Linear Formula [(C6H5)3P]2PdCl2
Infrared Spectrum Authentic
MDL Number MFCD00009593
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material Bis(triphenylphosphine)palladium(II) chloride
Assay 14.8 to 15.5% (Pd)
Assay Percent Range 98%
Molecular Formula C36H30Cl2P2Pd
Linear Formula [(C6H5)3P]2PdCl2
Formula Weight 701.89
MDL Number MFCD00009593
Synonym Dichlorobis(triphenylphosphine)palladium(II)
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents