Learn More
Bis(triphenylphosphine)nickel(II)chloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 217500500
Specifications
Bis(triphenylphosphine)nickel(II)chloride | |
Authentic | |
Gray to Green | |
50g | |
14264-16-5 | |
C36H32Cl2NiP2+2 | |
MFCD00009592 | |
bis triphenylphosphine dichloronickel ii | |
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl.Cl.[Ni+2] | |
656.193 | |
654.2 | |
98% |
97.5% min. | |
Glass bottle | |
>300.0°C | |
98% | |
98% | |
[(C6H5)3P]2NiCl2 | |
02,110; 09,147; 11,165; 16,118 | |
ZBRJXVVKPBZPAN-UHFFFAOYSA-N | |
nickel(2+);triphenylphosphane;dihydrochloride | |
131664342 | |
Crystals or Powder |
Chemical Identifiers
14264-16-5 | |
656.193 | |
ZBRJXVVKPBZPAN-UHFFFAOYSA-N | |
131664342 | |
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl.Cl.[Ni+2] |
C36H32Cl2NiP2+2 | |
MFCD00009592 | |
bis triphenylphosphine dichloronickel ii | |
nickel(2+);triphenylphosphane;dihydrochloride |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 238-154-8
RTECSNumber : QR6170000
TSCA : TSCA