missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(triphenylphosphine)nickel(II)chloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 217500100
Specifications
Bis(triphenylphosphine)nickel(II)chloride | |
Authentic | |
Gray to Green | |
10g | |
14264-16-5 | |
C36H30Cl2NiP2 | |
MFCD00009592 | |
bis triphenylphosphine dichloronickel ii | |
ZBRJXVVKPBZPAN-UHFFFAOYSA-N | |
nickel(2+);triphenylphosphane;dihydrochloride | |
131664342 | |
Crystals or Powder |
97.5% min. | |
Glass bottle | |
>300.0°C | |
98% | |
98% | |
[(C6H5)3P]2NiCl2 | |
02,110; 09,147; 11,165; 16,118 | |
Solubility in water: insoluble | |
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl.Cl.[Ni+2] | |
654.2 | |
654.2 | |
98% |
Chemical Identifiers
14264-16-5 | |
654.2 | |
ZBRJXVVKPBZPAN-UHFFFAOYSA-N | |
131664342 | |
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl.Cl.[Ni+2] |
C36H30Cl2NiP2 | |
MFCD00009592 | |
bis triphenylphosphine dichloronickel ii | |
nickel(2+);triphenylphosphane;dihydrochloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause cancer.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 238-154-8
RTECSNumber : QR6170000
TSCA : TSCA