Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(triphenylphosphine)iminium trifluoroacetate, 98%

Chemical Identifiers

Chemical Name or Material Bis(triphenylphosphine)iminium trifluoroacetate
CAS 116405-43-7
Formula Weight 782.65
MDL Number MFCD15146355
Synonym Bis(triphenylphosphoranylidene)ammonium trifluoroacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6155006

  SDS
alfa aesar™
H6155006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6155014

  SDS
alfa aesar™
H6155014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 782.65
Synonym Bis(triphenylphosphoranylidene)ammonium trifluoroacetate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Bis(triphenylphosphine)iminium trifluoroacetate
Assay 98%
Quantity 5g
MDL Number MFCD15146355
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 116405-43-7
Color White
Physical Form Solid

Applications
Bis(triphenylphosphine)iminium trifluoroacetate is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.