missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(triphenylphosphine)iminium trifluoroacetate, 98%

Manufacturer:  Alfa Aesar™ H6155014

 View more versions of this product

Catalog No. AAH6155014



Description

Description

Applications
Bis(triphenylphosphine)iminium trifluoroacetate is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

Bis(triphenylphosphine)iminium trifluoroacetate
25g
C38H30F3NO2P2
1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium 2,2,2-trifluoroacetate
C1=CC=C(C=C1)P(=N[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.C(=O)(C(F)(F)F)[O-]
651.606
782.65
98%
White
116405-43-7
MFCD15146355
NDLHFNFVQBXEPE-UHFFFAOYSA-M
2,2,2-trifluoroacetate;triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium
19800386
Solid

Chemical Identifiers

116405-43-7
651.606
NDLHFNFVQBXEPE-UHFFFAOYSA-M
19800386
C1=CC=C(C=C1)P(=N[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.C(=O)(C(F)(F)F)[O-]
C38H30F3NO2P2
MFCD15146355
1,1,1-triphenyl-n-triphenylphosphoranylidene phosphoraniminium 2,2,2-trifluoroacetate
2,2,2-trifluoroacetate;triphenyl-[(triphenyl-$l^{5}-phosphanylidene)amino]phosphanium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents