Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(triphenylphosphine)copper(I) borohydride

Chemical Identifiers

Chemical Name or Material Bis(triphenylphosphine)copper(I) borohydride
CAS 16903-61-0
Molecular Formula C36H34BCuP2
Linear Formula [(C6H5)3P]2CuBH4
Formula Weight 602.97
MDL Number MFCD00013219
UN Number UN2813
Synonym Bis(triphenylphosphine)copper(I) tetrahydridoborate; Tetrahydridoboratobis(triphenylphosphine)copper(I)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1373006

 
alfa aesar™
A1373006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1373014

 
alfa aesar™
A1373014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1373022

 
alfa aesar™
A1373022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 602.97
Chemical Name or Material Bis(triphenylphosphine)copper(I) borohydride
Molecular Formula C36H34BCuP2
Melting Point ≈175°C (dec.)
Linear Formula [(C6H5)3P]2CuBH4
MDL Number MFCD00013219
Quantity 5g
CAS 16903-61-0
UN Number UN2813
Synonym Bis(triphenylphosphine)copper(I) tetrahydridoborate; Tetrahydridoboratobis(triphenylphosphine)copper(I)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P231+P232-P261-P280-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P231+P232-P261-P280-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H261-H315-H319-H335
In contact with water releases flammable gas.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.