missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(trimethylsilyl)sulfide, tech.

Manufacturer:  Alfa Aesar™ 4481803

 View more versions of this product

Catalog No. AA4481803



Specifications

Specifications

Bis (trimethylsilyl)sulfide
26°C (79°F)
Ampule
Air Sensitive
Colorless
3385-94-2
[(CH3)3Si]2S
UN1993
bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, bis trimethylsilyl sulfide, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8
C[Si](C)(C)S[Si](C)(C)C
178.44
178.44
Technical
0.851g/mL
Pungent
1.4598
162°C to 166°C
1g
C6H18SSi2
MFCD00014851
1740046
RLECCBFNWDXKPK-UHFFFAOYSA-N
trimethyl(trimethylsilylsulfanyl)silane
76920
Liquid

Chemical Identifiers

3385-94-2
178.44
RLECCBFNWDXKPK-UHFFFAOYSA-N
76920
C[Si](C)(C)S[Si](C)(C)C
C6H18SSi2
MFCD00014851
bis trimethylsilyl sulfide, hexamethyldisilthiane, hexamethyldisilathiane, disilathiane, hexamethyl, bis trimethylsilyl sulfide, 1,1,1,3,3,3-hexamethyldisilathiane, disilthiane, hexamethyl, disilathiane, 1,1,1,3,3,3-hexamethyl, thiobis trimethylsilane, acmc-1ckj8
trimethyl(trimethylsilylsulfanyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

SDS Documents