Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(trimethylsilyl) sulfate, 97%

Chemical Identifiers

Chemical Name or Material Bis(trimethylsilyl) sulfate
CAS 18306-29-1
Molecular Formula C12H18O4SSi2
Linear Formula [(CH3)3SiO]2SO2
Formula Weight 242.44g/mol
MDL Number MFCD00008268
UN Number UN2925
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2360806

 
alfa aesar™
B23608-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2360814

 
alfa aesar™
B23608-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2360822

 
alfa aesar™
B23608-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 87° to 90°C (4mmHg)
Melting Point 44° to 47°C
Quantity 5g
Flash Point −2°C (27°F)
Chemical Name or Material Bis(trimethylsilyl) sulfate
CAS 18306-29-1
Assay 97%
Molecular Formula C12H18O4SSi2
Linear Formula [(CH3)3SiO]2SO2
Formula Weight 242.44g/mol
MDL Number MFCD00008268
UN Number UN2925
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.