Learn More
Alfa Aesar™ Bis(trimethylsilyl)methane, 98%
Manufacturer: Alfa Aesar™ A1566203
Specifications
Bis(trimethylsilyl)methane | |
C7H20Si2 | |
UN1993 | |
bis trimethylsilyl methane, methylenebis trimethylsilane, silane, methylenebis trimethyl, methylenebis trimethylsilane, trimethyl trimethylsilylmethyl silane, trimethyl trimethylsilyl methyl silane, 2,4-disilapentane, 2,2,4,4-tetramethyl, silane, methylenebis trimethyl, silane, 1,1'-methylenebis 1,1,1-trimethyl, silane, 1,1'-methylenebis 1,1,1-trimethyl | |
C[Si](C)(C)C[Si](C)(C)C | |
160.407 | |
160.41 | |
132°C to 133°C | |
1g |
2117-28-4 | |
MFCD00008275 | |
1734530 | |
GYIODRUWWNNGPI-UHFFFAOYSA-N | |
trimethyl(trimethylsilylmethyl)silane | |
75029 | |
98% | |
-71°C | |
12°C (53°F) |
Chemical Identifiers
2117-28-4 | |
160.407 | |
GYIODRUWWNNGPI-UHFFFAOYSA-N | |
75029 | |
C[Si](C)(C)C[Si](C)(C)C |
C7H20Si2 | |
MFCD00008275 | |
bis trimethylsilyl methane, methylenebis trimethylsilane, silane, methylenebis trimethyl, methylenebis trimethylsilane, trimethyl trimethylsilylmethyl silane, trimethyl trimethylsilyl methyl silane, 2,4-disilapentane, 2,2,4,4-tetramethyl, silane, methylenebis trimethyl, silane, 1,1'-methylenebis 1,1,1-trimethyl, silane, 1,1'-methylenebis 1,1,1-trimethyl | |
trimethyl(trimethylsilylmethyl)silane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : TSCA