missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(trimethylsilyl) malonate, 98%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 98%
Molecular Formula C9H20O4Si2
Linear Formula (CH3)3SiOC(O)CH2C(O)OSi(CH3)3
Formula Weight 248.43
MDL Number MFCD00009631
Chemical Name or Material Bis(trimethylsilyl) malonate
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC313300100 SDS Thermo Fisher Scientific
AC313300100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC313300500 SDS Thermo Fisher Scientific
AC313300500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

98%
(CH3)3SiOC(O)CH2C(O)OSi(CH3)3
MFCD00009631
≥96.0%
C9H20O4Si2
248.43
Bis(trimethylsilyl) malonate

Specifications

248.43
Liquid
0.972g/mL
Authentic
MFCD00009631
Glass bottle
C9H20O4Si2
≥96.0%
Clear colorless to straw
10g
(CH3)3SiOC(O)CH2C(O)OSi(CH3)3
98%
1.4150 to 1.4170 (20°C, 589nm)
18457-04-0
Bis(trimethylsilyl) malonate

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning