Learn More
Bis(trimethylsilyl)carbodiimide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 375130050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentSpecifications
Bis(trimethylsilyl)carbodiimide | |
-23°C | |
0.8210g/mL | |
42°C | |
97.5% min. (GC) | |
C7H18N2Si2 | |
MFCD00051538 | |
0.821 | |
Solubility in water: insoluble | |
C[Si](C)(C)N=C=N[Si](C)(C)C | |
186.4 | |
186.4 | |
Crystalline Powder, Crystals or Chunks |
1000-70-0 | |
Beige to Yellow or White | |
164°C | |
Authentic | |
Glass bottle | |
1.4330 to 1.435 | |
5 g | |
bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine | |
KSVMTHKYDGMXFJ-UHFFFAOYSA-N | |
N,N'-bis(trimethylsilyl)methanediimine | |
70473 | |
98% |
Chemical Identifiers
1000-70-0 | |
186.4 | |
KSVMTHKYDGMXFJ-UHFFFAOYSA-N | |
70473 | |
C[Si](C)(C)N=C=N[Si](C)(C)C |
C7H18N2Si2 | |
MFCD00051538 | |
bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine | |
N,N'-bis(trimethylsilyl)methanediimine |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 213-673-2
RUO – Research Use Only