missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(trimethylsilyl)carbodiimide, 97%

Manufacturer:  Alfa Aesar™ A1167003

 View more versions of this product

Catalog No. AAA1167003



Specifications

Specifications

Bis (trimethylsilyl)carbodiimide
Mild
-23°C
1000-70-0
(CH3)3SiN=C=NSi(CH3)3
UN1993
bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
C[Si](C)(C)N=C=N[Si](C)(C)C
186.405
186.41
42°C (107°F)
163°C to 164°C
1g
C7H18N2Si2
MFCD00051538
2042082
KSVMTHKYDGMXFJ-UHFFFAOYSA-N
N,N'-bis(trimethylsilyl)methanediimine
70473
97%

Chemical Identifiers

1000-70-0
186.405
KSVMTHKYDGMXFJ-UHFFFAOYSA-N
70473
C[Si](C)(C)N=C=N[Si](C)(C)C
C7H18N2Si2
MFCD00051538
bis trimethylsilyl carbodiimide, carbodiimide, bis trimethylsilyl, n,n'-methanediylidenebis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, 1,3-bis trimethylsilyl carbodiimide, n,n'-bis trimethylsilyl carbodiimide, n,n'-methane tetraylbis 1,1,1-trimethylsilanamine, silanamine, n,n'-methanetetraylbis 1,1,1-trimethyl, carbodiimide, bis trimethylsilyl-7ci,8ci, n,n'-methanetetraylbis 1,1,1-trimethylsilylamine
N,N'-bis(trimethylsilyl)methanediimine
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280h-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : VV1302500

TSCA : TSCA

SDS Documents