Please login to your online account to display your discounted pricing

[Bis(trifluoroacetoxy)iodo]benzene, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC213670500

 View more versions of this product

Catalog No. AC213670500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 430.03
Physical Form Crystalline Powder
Quantity 50g
Color White to pale yellow
Merck Index 15, 7406
Melting Point 121° to 125°C
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula (CF3CO2)2IC6H5
Assay Percent Range 98%
CAS 2712-78-9
MDL Number MFCD00009672
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
CAS Min % 97.5
Packaging Glass bottle
Name Note 98%
Chemical Name or Material [Bis(trifluoroacetoxy)iodo]benzene
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C10H5F6IO4

Specifications

Chemical Name or Material [Bis(trifluoroacetoxy)iodo]benzene
Name Note 98%
CAS 2712-78-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H5F6IO4
Linear Formula (CF3CO2)2IC6H5
Formula Weight 430.03
MDL Number MFCD00009672
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents