missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[Bis(trifluoroacetoxy)iodo]benzene, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 213670100

 View more versions of this product

Catalog No. AC213670100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

oxidation reagent
Specifications

Specifications

[Bis(trifluoroacetoxy)iodo]benzene
2712-78-9
100.0
97.5% min. (Iodometry)
C10H5F6IO4
MFCD00009672
09,54
bis trifluoroacetoxy iodo benzene, pifa, unii-659sfv27xs, bis-trifluoroacetoxyiodobenzene, phenylbis trifluoroacetato-o iodine, bis i,i-trifluoroacetoxy iodobenzene, iodine, phenylbis trifluoroacetato-o, bis trifluoracetoxy-iodobenzene, bis trifluoroacetoxy phenyl iodane
PEZNEXFPRSOYPL-UHFFFAOYSA-N
[phenyl-(2,2,2-trifluoroacetyl)oxy-$l^{3}-iodanyl] 2,2,2-trifluoroacetate
102317
98%
98%
97.5
Authentic
Glass bottle
(CF3CO2)2IC6H5
10g
15, 7406
Solubility in water: insoluble. Other solubilities: soluble in organic solvent
C1=CC=C(C=C1)I(OC(=O)C(F)(F)F)OC(=O)C(F)(F)F
430.03
430.03

Chemical Identifiers

2712-78-9
430.03
PEZNEXFPRSOYPL-UHFFFAOYSA-N
102317
C1=CC=C(C=C1)I(OC(=O)C(F)(F)F)OC(=O)C(F)(F)F
C10H5F6IO4
MFCD00009672
bis trifluoroacetoxy iodo benzene, pifa, unii-659sfv27xs, bis-trifluoroacetoxyiodobenzene, phenylbis trifluoroacetato-o iodine, bis i,i-trifluoroacetoxy iodobenzene, iodine, phenylbis trifluoroacetato-o, bis trifluoracetoxy-iodobenzene, bis trifluoroacetoxy phenyl iodane
[phenyl-(2,2,2-trifluoroacetyl)oxy-$l^{3}-iodanyl] 2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-308-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only