missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(triethylphosphine)palladium(II) chloride, ACROS Organics™

Chemical Identifiers

CAS 28425-04-9
Molecular Formula C12H30Cl2P2Pd
Formula Weight 413.64
MDL Number MFCD00191831
Chemical Name or Material Bis(triethylphosphine)palladium(II) chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC377970010 SDS Thermo Fisher Scientific
AC377970010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC377972500 SDS Thermo Fisher Scientific
AC377972500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

28425-04-9
413.64
Bis(triethylphosphine)palladium(II) chloride
C12H30Cl2P2Pd
MFCD00191831

Specifications

28425-04-9
Glass bottle
Powder
413.64
Yellow
Bis(triethylphosphine)palladium(II) chloride
Authentic
24.9% min.
C12H30Cl2P2Pd
MFCD00191831
1g

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning