missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(tricarbonylcyclopentadienylmolybdenum)

Manufacturer:  Alfa Aesar™ 4812106

 View more versions of this product

Catalog No. AA4812106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Bis (tricarbonylcyclopentadienylmolybdenum)
C16H2Mo2O6
UN3466
180°C (decomposition)
5g
490.13
12091-64-4
MFCD00009947
Cyclopentadienylmolybdenum tricarbonyl dimer; Tricarbonylcyclopentadienylmolybdenum dimer
AYBJYGWNVNZERR-UHFFFAOYSA-N
482.09
Crystalline

Chemical Identifiers

12091-64-4
482.09
AYBJYGWNVNZERR-UHFFFAOYSA-N
C16H2Mo2O6
MFCD00009947
Cyclopentadienylmolybdenum tricarbonyl dimer; Tricarbonylcyclopentadienylmolybdenum dimer
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.

Documents