missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(tributyltin), TCI America™

$77.92 - $235.28

Chemical Identifiers

CAS 813-19-4
Molecular Formula C24H54Sn2
Molecular Weight (g/mol) 580.116
MDL Number MFCD00009417
InChI Key REDSKZBUUUQMSK-UHFFFAOYSA-N
Synonym hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane
PubChem CID 6327815
IUPAC Name tributyltin
SMILES CCCC[Sn](CCCC)CCCC.CCCC[Sn](CCCC)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B18325G View Documents TCI America
B18325G
5g Each for $77.92
 
B183225G View Documents TCI America
B183225G
25g Each for $235.28
 
Specifications

Chemical Identifiers

813-19-4
580.116
REDSKZBUUUQMSK-UHFFFAOYSA-N
6327815
CCCC[Sn](CCCC)CCCC.CCCC[Sn](CCCC)CCCC
C24H54Sn2
MFCD00009417
hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane
tributyltin

Specifications

194°C
5g
C24H54Sn2
2788
REDSKZBUUUQMSK-UHFFFAOYSA-N
tributyltin
6327815
Clear Liquid at 20°C
Bis(tributyltin)
Colorless
813-19-4
MFCD00009417
hexabutylditin, hexabutyldistannane, bis, distannane, hexabutyl, tin, hexabutyldi, stannane, hexabutyldi, hexa-n-butylditin, 1,1,1,2,2,2-hexabutyldistannane, hexabutyidistannane, hexabutyl distannane
CCCC[Sn](CCCC)CCCC.CCCC[Sn](CCCC)CCCC
580.116
580.12
≥93.0% (GC)
Safety and Handling

Safety and Handling

TSCA : Yes