missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(tributylstannyl)acetylene, TCI America™

$123.75 - $351.39

Chemical Identifiers

CAS 994-71-8
Molecular Formula C26H54Sn2
Molecular Weight (g/mol) 604.138
MDL Number MFCD00010239
InChI Key CAUONNQGFXOEMY-UHFFFAOYSA-N
Synonym bis tributylstannyl acetylene, 1,2-bis tributylstannyl ethyne, bis tri-n-butylstannyl acetylene, bis tri-n-butylstannyl ethyne, bis tributylstannyl acetylene, tributyl 2-tributylstannylethynyl stannane, tributyl tributylstannyl ethynyl stannane #, tributyl 2-tributylstannyl ethynyl stannane
PubChem CID 305981
IUPAC Name tributyl(2-tributylstannylethynyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B19741G View Documents TCI America
B19741G
1g Each for $123.75
 
B19745G View Documents TCI America
B19745G
5g Each for $351.39
 
Specifications

Chemical Identifiers

994-71-8
604.138
CAUONNQGFXOEMY-UHFFFAOYSA-N
305981
CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC
C26H54Sn2
MFCD00010239
bis tributylstannyl acetylene, 1,2-bis tributylstannyl ethyne, bis tri-n-butylstannyl acetylene, bis tri-n-butylstannyl ethyne, bis tributylstannyl acetylene, tributyl 2-tributylstannylethynyl stannane, tributyl tributylstannyl ethynyl stannane #, tributyl 2-tributylstannyl ethynyl stannane
tributyl(2-tributylstannylethynyl)stannane

Specifications

Yellow
994-71-8
MFCD00010239
bis tributylstannyl acetylene, 1,2-bis tributylstannyl ethyne, bis tri-n-butylstannyl acetylene, bis tri-n-butylstannyl ethyne, bis tributylstannyl acetylene, tributyl 2-tributylstannylethynyl stannane, tributyl tributylstannyl ethynyl stannane #, tributyl 2-tributylstannyl ethynyl stannane
CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC
604.138
604.14
≥95.0% (GC)
1g
C26H54Sn2
2788
CAUONNQGFXOEMY-UHFFFAOYSA-N
tributyl(2-tributylstannylethynyl)stannane
305981
Clear Liquid at 20°C
Bis(tributylstannyl)acetylene
Safety and Handling

Safety and Handling

TSCA : No