missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(tributylstannyl)acetylene, 97%, ACROS Organics™

$342.64 - $342.64

Chemical Identifiers

CAS 994-71-8
Molecular Formula C26H54Sn2
Molecular Weight (g/mol) 604.13
MDL Number MFCD00010239
InChI Key CAUONNQGFXOEMY-UHFFFAOYSA-N
Synonym 1,2-bis tributylstannyl ethyne, bis tri-n-butylstannyl acetylene, bis tri-n-butylstannyl ethyne, bis tributylstannyl acetylene, tributyl 2-tributylstannyl ethynyl stannane, tributyl 2-tributylstannylethynyl stannane, tributyl tributylstannyl ethynyl stannane #
PubChem CID 305981
IUPAC Name tributyl(2-tributylstannylethynyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC369580050
SDS View Documents
Thermo Fisher Scientific
AC369580050
5g Glass bottle
Each for $342.64
 
Description

Description

Specifications

Chemical Identifiers

994-71-8
604.13
CAUONNQGFXOEMY-UHFFFAOYSA-N
305981
CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC
C26H54Sn2
MFCD00010239
1,2-bis tributylstannyl ethyne, bis tri-n-butylstannyl acetylene, bis tri-n-butylstannyl ethyne, bis tributylstannyl acetylene, tributyl 2-tributylstannyl ethynyl stannane, tributyl 2-tributylstannylethynyl stannane, tributyl tributylstannyl ethynyl stannane #
tributyl(2-tributylstannylethynyl)stannane

Specifications

1.1470g/mL
Authentic
1.4920 to 1.4940
Colorless
994-71-8
C26H54Sn2
MFCD00010239
CAUONNQGFXOEMY-UHFFFAOYSA-N
tributyl(2-tributylstannylethynyl)stannane
305981
Liquid
Bis(tributylstannyl)acetylene
>110°C
Glass bottle
1.147
5g
96% min. (GC)
[CH3(CH2)3]3SnC≡CSn[(CH2)3CH3]3
1,2-bis tributylstannyl ethyne, bis tri-n-butylstannyl acetylene, bis tri-n-butylstannyl ethyne, bis tributylstannyl acetylene, tributyl 2-tributylstannyl ethynyl stannane, tributyl 2-tributylstannylethynyl stannane, tributyl tributylstannyl ethynyl stannane #
CCCC[Sn](CCCC)(CCCC)C#C[Sn](CCCC)(CCCC)CCCC
604.13
604.13
97%
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with

GHS P Statement
Take off contaminated clothing and wash before reuse.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash wi

GHS Signal Word: Danger

SDS