Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(tri-n-butyltin) sulfate, 97%

Chemical Identifiers

Chemical Name or Material Bis(tri-n-butyltin) sulfate
CAS 26377-04-8
Formula Weight 676.14
MDL Number MFCD00185908
UN Number UN3146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3962409

 
alfa aesar™
3962409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3962418

 
alfa aesar™
3962418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 676.14
Chemical Name or Material Bis(tri-n-butyltin) sulfate
Physical Form Powder
Assay 97%
MDL Number MFCD00185908
CAS 26377-04-8
Quantity 10g
UN Number UN3146
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.