Please login to your online account to display your discounted pricing

Bis(tri-n-butyltin) Oxide, Stabilized 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(tri-n-butyltin) oxide
Name Note 96%
CAS 56-35-9
Assay ≥95% (ex Sn)
Assay Percent Range 96%
Molecular Formula C24H54OSn2
Linear Formula [CH3(CH2)3]3SnOSn[(CH2)3CH3]3
Formula Weight 596.11
MDL Number MFCD00009418
Synonym HBD
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC106510025

  SDS
thermo fisher scientific
AC106510025
Glass bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC106515000

  SDS
thermo fisher scientific
106515000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95% (ex Sn)
Physical Form Liquid
Formula Weight 596.11
Quantity 2.5L
Color Clear colorless to faintly yellow
EINECS Number 200-268-0
TSCA TSCA
Density 1.1700g/mL
RTECS Number JN8750000
Fieser 06,56; 07,26; 08,43; 09,53; 11,62; 13,41; 16,227
Name Note 96%
Molecular Formula C24H54OSn2
Linear Formula [CH3(CH2)3]3SnOSn[(CH2)3CH3]3
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Bis(tri-n-butyltin) oxide
Packaging Glass bottle
Assay Percent Range 96%
Refractive Index 1.4854 to 1.4874 (20°C, 589nm)
MDL Number MFCD00009418
CAS 56-35-9
Synonym HBD
Infrared Spectrum Authentic
Stabilizer 0.1% lactic acid
Chlorine (Cl) 0.1% max.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.