Please login to your online account to display your discounted pricing

Bis(tri-n-butyltin) Oxide, Stabilized 96%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  106515000

 View more versions of this product

Catalog No. AC106515000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95% (ex Sn)
Physical Form Liquid
Formula Weight 596.11
Quantity 500mL
Color Clear colorless to faintly yellow
EINECS Number 200-268-0
TSCA TSCA
RTECS Number JN8750000
Fieser 06,56; 07,26; 08,43; 09,53; 11,62; 13,41; 16,227
Chemical Name or Material Bis(tri-n-butyltin) oxide
Name Note 96%
Packaging Glass bottle
Assay Percent Range 96%
Refractive Index 1.4854 to 1.4874 (20°C, 589nm)
Molecular Formula C24H54OSn2
Linear Formula [CH3(CH2)3]3SnOSn[(CH2)3CH3]3
MDL Number MFCD00009418
CAS 56-35-9
Chlorine (Cl) 0.1% max.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Infrared Spectrum Authentic
Stabilizer 0.1% lactic acid
Synonym HBD
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Bis(tri-n-butyltin) oxide
Name Note 96%
CAS 56-35-9
Assay ≥95% (ex Sn)
Assay Percent Range 96%
Molecular Formula C24H54OSn2
Linear Formula [CH3(CH2)3]3SnOSn[(CH2)3CH3]3
Formula Weight 596.11
MDL Number MFCD00009418
Synonym HBD
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents