Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(tetramethylcyclopentadienyl)manganese(II), 98+%

$136.97 - $476.14

Chemical Identifiers

Chemical Name or Material Bis(tetramethylcyclopentadienyl)manganese(II)
CAS 101932-75-6
Molecular Formula C18H26Mn
Formula Weight 297.35
MDL Number MFCD01862454
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2702703

  SDS
alfa aesar™
H2702703
1g Each for $136.97

AAH2702706

  SDS
alfa aesar™
H2702706
5g Each for $476.14
Description & Specifications

Specifications

Formula Weight 297.35
Chemical Name or Material Bis(tetramethylcyclopentadienyl)manganese(II)
Assay ≥98%
Molecular Formula C18H26Mn
MDL Number MFCD01862454
CAS 101932-75-6
Melting Point 98° to 112°C
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C