Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(tetramethylcyclopentadienyl)chromium(II), 98+%

$123.59 - $441.77

Chemical Identifiers

Chemical Name or Material Bis(tetramethylcyclopentadienyl)chromium(II)
CAS 82066-37-3
Molecular Formula C18H26Cr
Formula Weight 294.41
MDL Number MFCD01862451
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2739603

  SDS
alfa aesar™
H2739603
1g Each for $123.59

AAH2739606

  SDS
alfa aesar™
H2739606
5g Each for $441.77
Description & Specifications

Specifications

Formula Weight 294.41
Chemical Name or Material Bis(tetramethylcyclopentadienyl)chromium(II)
Assay ≥98%
Molecular Formula C18H26Cr
MDL Number MFCD01862451
CAS 82066-37-3
Melting Point 106° to 111°C
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.

2° to 8°C