missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L17046MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bis(tetraethylammonium) bis(2-thioxo-1,3-dithiole-4,5-dithiolato)zincate | |
202°C to 206°C | |
MFCD00623815 | |
4900651 | |
bis tetraethylammonium bis 2-thioxo-1,3-dithiole-4,5-dithiolato zincate | |
CC[N+](CC)(CC)CC.CC[N+](CC)(CC)CC.C1(=C(SC(=S)S1)[S-])[S-].C1(=C(SC(=S)S1)[S-])[S-].[Zn+2] | |
718.556 | |
718.6 |
72022-68-5 | |
C22H40N2S10Zn | |
250 mg | |
Light sensitive | |
TXRIVAQWJHZFOT-UHFFFAOYSA-J | |
zinc;2-sulfanylidene-1,3-dithiole-4,5-dithiolate;tetraethylazanium | |
15762389 | |
97% |
Chemical Identifiers
72022-68-5 | |
718.556 | |
TXRIVAQWJHZFOT-UHFFFAOYSA-J | |
15762389 | |
CC[N+](CC)(CC)CC.CC[N+](CC)(CC)CC.C1(=C(SC(=S)S1)[S-])[S-].C1(=C(SC(=S)S1)[S-])[S-].[Zn+2] |
C22H40N2S10Zn | |
MFCD00623815 | |
bis tetraethylammonium bis 2-thioxo-1,3-dithiole-4,5-dithiolato zincate | |
zinc;2-sulfanylidene-1,3-dithiole-4,5-dithiolate;tetraethylazanium |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only