missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(tetrabutylammonium) Bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc Complex, TCI America™

[Organic Electronic Material]

$334.59 - $1,145.86

Chemical Identifiers

CAS 68449-38-7
Molecular Formula C38H72N2S10Zn
Molecular Weight (g/mol) 942.99
MDL Number MFCD00060267
InChI Key ZBBYMGVOKYWSPO-UHFFFAOYSA-J
Synonym BTBA-BDTD Zn Complex
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B12215G View Documents TCI America
B12215G
5g Each for $334.59
 
B122125G View Documents TCI America
B122125G
25g Each for $1,145.86
 
Specifications

Chemical Identifiers

68449-38-7
942.99
ZBBYMGVOKYWSPO-UHFFFAOYSA-J
C38H72N2S10Zn
MFCD00060267
BTBA-BDTD Zn Complex

Specifications

Purple-Red
5g
C38H72N2S10Zn
BTBA-BDTD Zn Complex
942.99
Crystal-Powder at 20°C
Bis(tetrabutylammonium) Bis(1,3-dithiole-2-thione-4,5-dithiolato)zinc Complex [Organic Electronic Material]
174°C
68449-38-7
MFCD00060267
ZBBYMGVOKYWSPO-UHFFFAOYSA-J
942.99
≥98.0% (N,T)
Safety and Handling

Safety and Handling

TSCA : No