Please login to your online account to display your discounted pricing

Bis(tert-butylimido)osmium dioxide, ACROS Organics™


Quantity 50mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC375710500

 View more versions of this product

Catalog No. AC375710500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 364.44
Quantity 50mg
Color Dark brown
Chemical Name or Material Bis(tert-butylimido)osmium dioxide
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C8H18N2O2Os
MDL Number MFCD07368649
CAS 63174-13-0
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Bis(tert-butylimido)osmium dioxide
CAS 63174-13-0
Molecular Formula C8H18N2O2Os
Formula Weight 364.44
MDL Number MFCD07368649
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.

SDS & Safety Documents