Bis(tert-butylimido)osmium dioxide, ACROS Organics™
Manufacturer:  Thermo Fisher Scientific AC375710100
 View more versions of this product
Catalog No. AC375710100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
Powder
10mg
Bis(tert-butylimido)osmium dioxide
C8H18N2O2Os
Glass bottle
364.44
Dark brown
Authentic
63174-13-0
MFCD07368649
Chemical Identifiers
63174-13-0
364.44
Bis(tert-butylimido)osmium dioxide
C8H18N2O2Os
MFCD07368649
Safety and Handling
SDS & Safety Documents
GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger