Please login to your online account to display your discounted pricing

Bis(tert-butylimido)osmium dioxide, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(tert-butylimido)osmium dioxide
CAS 63174-13-0
Molecular Formula C8H18N2O2Os
Formula Weight 364.44
MDL Number MFCD07368649
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375710100

  SDS
thermo fisher scientific
AC375710100
Glass bottle 10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC375710500

  SDS
thermo fisher scientific
AC375710500
Glass bottle 50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 364.44
Quantity 10mg
Color Dark brown
Chemical Name or Material Bis(tert-butylimido)osmium dioxide
Packaging Glass bottle
Molecular Formula C8H18N2O2Os
MDL Number MFCD07368649
CAS 63174-13-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.