Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(tert-butylcyclopentadienyl)zirconium(IV) dichloride, 97%

$170.27 - $524.35

Chemical Identifiers

Chemical Name or Material Bis(tert-butylcyclopentadienyl)zirconium(IV) dichloride
CAS 32876-92-9
Molecular Formula C18H26Cl2Zr
Formula Weight 404.53
MDL Number MFCD01073785
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2689203

  SDS
alfa aesar™
H2689203
1g Each for $170.27

AAH2689206

  SDS
alfa aesar™
H2689206
5g Each for $524.35
Description & Specifications

Specifications

Formula Weight 404.53
Melting Point 192° to 194°C
Quantity 1g
Molecular Formula C18H26Cl2Zr
CAS 32876-92-9
Chemical Name or Material Bis(tert-butylcyclopentadienyl)zirconium(IV) dichloride
MDL Number MFCD01073785
Assay 97%