Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(tert-butylcyclopentadienyl)hafnium(IV) dichloride, 98+%

$166.24 - $601.86

Chemical Identifiers

Chemical Name or Material Bis(tert-butylcyclopentadienyl)hafnium(IV) dichloride
CAS 33010-55-8
Molecular Formula C18H26Cl2Hf
Formula Weight 491.8
MDL Number MFCD01073803
Synonym 1,1'-Di-tert-butylhafnocene dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2763803

 
alfa aesar™
H2763803
1g Each for $166.24

AAH2763806

 
alfa aesar™
H2763806
5g Each for $601.86
Description & Specifications

Specifications

Formula Weight 491.8
Chemical Name or Material Bis(tert-butylcyclopentadienyl)hafnium(IV) dichloride
Assay ≥98%
Molecular Formula C18H26Cl2Hf
MDL Number MFCD01073803
CAS 33010-55-8
Synonym 1,1'-Di-tert-butylhafnocene dichloride
Melting Point 195°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.