Learn More
(-)-Bis[(S)-1-phenylethyl]amine, ChiPros™, 99%, ee 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2710614
Description
(-)-Bis[(S)-1-phenylethyl]amine is used as a chiral resolution reagent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(-)-Bis[(S)-1-phenylethyl]amine is used as a chiral resolution reagent in organic synthesis.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
(-)-Bis[(S)-1-phenylethyl]amine | |
98°C (0.3 mmHg) | |
C16H19N | |
25 g | |
--bis s-1-phenylethyl amine, s-bis s-1-phenylethyl amine, unii-5d97t2y7wu, bis s-1-phenylethyl amine, 1s-1-phenyl-n-1s-1-phenylethyl ethanamine, bis 1s-1-phenylethyl amine, s,s-di 1-phenylethyl amine | |
NXLACVVNHYIYJN-KBPBESRZSA-N | |
(1S)-1-phenyl-N-[(1S)-1-phenylethyl]ethanamine | |
6994958 | |
99% |
56210-72-1 | |
ee ≥98% | |
MFCD00243087 | |
Air Sensitive | |
Immiscible with water. | |
CC(C1=CC=CC=C1)NC(C)C2=CC=CC=C2 | |
225.335 | |
225.34 |
Chemical Identifiers
56210-72-1 | |
225.335 | |
NXLACVVNHYIYJN-KBPBESRZSA-N | |
6994958 | |
CC(C1=CC=CC=C1)NC(C)C2=CC=CC=C2 |
C16H19N | |
MFCD00243087 | |
--bis s-1-phenylethyl amine, s-bis s-1-phenylethyl amine, unii-5d97t2y7wu, bis s-1-phenylethyl amine, 1s-1-phenyl-n-1s-1-phenylethyl ethanamine, bis 1s-1-phenylethyl amine, s,s-di 1-phenylethyl amine | |
(1S)-1-phenyl-N-[(1S)-1-phenylethyl]ethanamine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only