Please login to your online account to display your discounted pricing

Bis(pinacolato)diboron, 98%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  330572500

 View more versions of this product

Catalog No. AC330572500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder or Platelets
Formula Weight 253.9
Quantity 250mg
Color White
Melting Point 135° to 140°C
Chemical Name or Material Bis(pinacolato)diboron
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C12H24B2O4
MDL Number MFCD00799570
CAS 73183-34-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Bis(pinacolato)diboron
CAS 73183-34-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H24B2O4
Formula Weight 253.9
MDL Number MFCD00799570
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents