Please login to your online account to display your discounted pricing

Bis(phenylsulfonyl)sulfide, 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC300830050

 View more versions of this product

Catalog No. AC300830050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder and/or Crystals
Assay ≥95% (HPLC)
Formula Weight 314.41
Quantity 5g
Color Off-white to pink
Melting Point 129° to 134°C
CAS 4388-22-1
Molecular Formula C12H10O4S3
Assay Percent Range 96%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Bis(phenylsulfonyl)sulfide

Specifications

Chemical Name or Material Bis(phenylsulfonyl)sulfide
Assay ≥95% (HPLC)
Assay Percent Range 96%
Molecular Formula C12H10O4S3
Formula Weight 314.41