Please login to your online account to display your discounted pricing

Bis(pentamethylcyclopentadienyl)titanium dichloride 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(pentamethylcyclopentadienyl)titanium dichloride
CAS 11136-36-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C20H30Cl2Ti
Formula Weight 389.24
MDL Number MFCD00049153
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383430010

  SDS
thermo fisher scientific
AC383430010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC383430050

  SDS
thermo fisher scientific
AC383430050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 389.24
Physical Form Crystals or Needles
Quantity 1g
Color Red or brown
Chemical Name or Material Bis(pentamethylcyclopentadienyl)titanium dichloride
Assay Percent Range 99%
Molecular Formula C20H30Cl2Ti
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00049153
CAS 11136-36-0
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.