Please login to your online account to display your discounted pricing

Bis(pentamethylcyclopentadienyl)manganese, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  35326-0050

 View more versions of this product

Catalog No. AC353260050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 325.39
Physical Form Crystalline Powder
Quantity 5g
Color Red-brown
Health Hazard 2 AIR SENSITIVE
Infrared Spectrum Authentic
Health Hazard 1 MOISTURE SENSITIVE
CAS 67506-86-9
Assay Percent Range 98%
Chemical Name or Material Bis(pentamethylcyclopentadienyl)manganese
Molecular Formula C20H30Mn
MDL Number MFCD00058844
NMR 97.5% min.
Packaging Glass bottle

Specifications

Chemical Name or Material Bis(pentamethylcyclopentadienyl)manganese
Assay Percent Range 98%
Molecular Formula C20H30Mn
Formula Weight 325.39
MDL Number MFCD00058844
Accessories

MOISTURE SENSITIVE

AIR SENSITIVE

SDS & Safety Documents