Please login to your online account to display your discounted pricing

Bis(pentamethylcyclopentadienyl)manganese, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(pentamethylcyclopentadienyl)manganese
Assay Percent Range 98%
Molecular Formula C20H30Mn
Formula Weight 325.39
MDL Number MFCD00058844
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC353260050

  SDS
thermo fisher scientific
35326-0050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC353260010

  SDS
thermo fisher scientific
35326-0010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 325.39
Quantity 5g
Color Red-brown
Infrared Spectrum Authentic
CAS 67506-86-9
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 AIR SENSITIVE
Chemical Name or Material Bis(pentamethylcyclopentadienyl)manganese
NMR 97.5% min.
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C20H30Mn
MDL Number MFCD00058844
Safety and Handling

MOISTURE SENSITIVE

AIR SENSITIVE