Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(pentamethylcyclopentadienyl)magnesium, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Bis(pentamethylcyclopentadienyl)magnesium
CAS 74507-64-5
Formula Weight 294.77
MDL Number MFCD00151387
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3859804

  SDS
alfa aesar™
3859804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3859802

  SDS
alfa aesar™
3859802
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3859809

  SDS
alfa aesar™
3859809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 294.77
Packaging Ampule
Chemical Name or Material Bis(pentamethylcyclopentadienyl)magnesium
Physical Form Crystalline
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00151387
CAS 74507-64-5
UN Number UN3181
Quantity 2g
Health Hazard 1 Danger
Sensitivity Air & Moisture Sensitive
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.