Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride, 98+%

$113.01 - $364.64

Chemical Identifiers

Chemical Name or Material Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride
CAS 85959-83-7
Molecular Formula C20H30Cl2Hf
Formula Weight 519.86
MDL Number MFCD00058887
Synonym Decamethylhafnocene dichloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2691203

  SDS
alfa aesar™
H2691203
1g Each for $113.01

AAH2691206

  SDS
alfa aesar™
H2691206
5g Each for $364.64
Description & Specifications

Specifications

Formula Weight 519.86
Melting Point 300°C
Molecular Formula C20H30Cl2Hf
Synonym Decamethylhafnocene dichloride
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride
MDL Number MFCD00058887
CAS 85959-83-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.