Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct

Chemical Identifiers

Chemical Name or Material Bis (pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct
CAS 312739-90-5
Molecular Formula C20H30Ba·C4H10O2
Formula Weight 479.92
MDL Number MFCD02684523
UN Number UN1564
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4363903

 
alfa aesar™
4363903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4363906

 
alfa aesar™
4363906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43639MD

 
alfa aesar™
43639MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 479.92
Packaging Ampule
Chemical Name or Material Bis (pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct
Physical Form Powder
Molecular Formula C20H30Ba·C4H10O2
MDL Number MFCD02684523
CAS 312739-90-5
UN Number UN1564
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

  • Ampouled under argon