Alfa Aesar™ Bis(pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct
Manufacturer:  Alfa Aesar™ 43639MD
 View more versions of this product
Catalog No. AA43639MD
Description
Description
  • Ampouled under argon
Specifications
Specifications
Bis (pentamethylcyclopentadienyl)barium 1,2-dimethoxyethane adduct
250mg
C20H30Ba
UN1564
ZVWNPBSWYPWMIF-UHFFFAOYSA-N
barium(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene
21946354
Powder
Ampule
312739-90-5
MFCD02684523
acmc-1aijw
CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ba+2]
407.787
479.92
Chemical Identifiers
312739-90-5
407.787
ZVWNPBSWYPWMIF-UHFFFAOYSA-N
21946354
CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ba+2]
C20H30Ba
MFCD02684523
acmc-1aijw
barium(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene
Safety and Handling
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
storageNote1 :
  • Ampouled under argon
Documents