Please login to your online account to display your discounted pricing

Bis(pentafluorophenyl) carbonate, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC374670010

 View more versions of this product

Catalog No. AC374670010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 394.12
Physical Form Crystalline Powder and Chunks
Quantity 1g
Color White
Melting Point 48° to 51°C
Infrared Spectrum Authentic
CAS 59483-84-0
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C13F10O3
Chemical Name or Material Bis(pentafluorophenyl) carbonate
MDL Number MFCD00368353

Specifications

Chemical Name or Material Bis(pentafluorophenyl) carbonate
CAS 59483-84-0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13F10O3
Formula Weight 394.12
MDL Number MFCD00368353