Please login to your online account to display your discounted pricing

Bis(pentafluorophenyl) carbonate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(pentafluorophenyl) carbonate
CAS 59483-84-0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13F10O3
Formula Weight 394.12
MDL Number MFCD00368353
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC374670050

  SDS
thermo fisher scientific
AC374670050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC374670010

  SDS
thermo fisher scientific
AC374670010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder and Chunks
Formula Weight 394.12
Quantity 5g
Color White
Melting Point 48° to 51°C
Infrared Spectrum Authentic
Molecular Formula C13F10O3
Chemical Name or Material Bis(pentafluorophenyl) carbonate
Assay Percent Range 98%
MDL Number MFCD00368353
CAS 59483-84-0
Packaging Glass bottle

Coupling reagent