missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(pentafluorophenyl) carbonate, 98+%

Manufacturer:  Alfa Aesar™ H5488014

 View more versions of this product

Catalog No. AAH5488014



Description

Description

Applications
Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.

Solubility
Soluble in chloroform, ethyl acetate and methylene chloride.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

Bis(pentafluorophenyl) carbonate
47°C to 50°C
59483-84-0
(C6F5O)2CO
2029727
Soluble in chloroform,ethyl acetate and methylene chloride.
C1(=C(C(=C(C(=C1F)F)F)F)F)OC(=O)OC2=C(C(=C(C(=C2F)F)F)F)F
394.124
394.12
110°C (230°F)
25g
C13F10O3
MFCD00368353
acmc-209mds
IOVVFSGCNWQFQT-UHFFFAOYSA-N
bis(2,3,4,5,6-pentafluorophenyl) carbonate
2734833
≥98%

Chemical Identifiers

59483-84-0
394.124
IOVVFSGCNWQFQT-UHFFFAOYSA-N
2734833
C1(=C(C(=C(C(=C1F)F)F)F)F)OC(=O)OC2=C(C(=C(C(=C2F)F)F)F)F
C13F10O3
MFCD00368353
acmc-209mds
bis(2,3,4,5,6-pentafluorophenyl) carbonate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents