Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(pentabromophenyl) ether, 99%

$8.45 - $8.45

Chemical Identifiers

Chemical Name or Material Bis(pentabromophenyl) ether
CAS 1163-19-5
Molecular Formula C12Br10O
Formula Weight 959.17
MDL Number MFCD00000059
UN Number UN3152
Beilstein 2188438
Synonym Decabromodiphenyl ether; Pentabromophenyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0465614

  SDS
alfa aesar™
L0465614
25g Each for $8.45
Description & Specifications

Specifications

Formula Weight 959.17
Chemical Name or Material Bis(pentabromophenyl) ether
MDL Number MFCD00000059
CAS 1163-19-5
TSCA TSCA
Assay 99%
Molecular Formula C12Br10O
UN Number UN3152
Beilstein 2188438
Synonym Decabromodiphenyl ether; Pentabromophenyl ether
Melting Point 304° to 308°C
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H351-H302-H312
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
RTECS Number KN3525000