missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(norbornadiene)rhodium(I) tetrafluoroborate

Manufacturer:  Alfa Aesar™ 4403606

 View more versions of this product

Catalog No. AA4403606



Specifications

Specifications

Bis (norbornadiene)rhodium (I) tetrafluoroborate
5g
C14H16BF4Rh-
bis bicyclo 2.2.1 hepta-2,5-diene rhodium tetrafluoroborate
ZWPWNBFOFVFMPP-UHFFFAOYSA-N
bicyclo[2.2.1]hepta-2,5-diene;rhodium;tetrafluoroborate
10915722
Powder
Air and moisture sensitive
36620-11-8
MFCD00671775
Fairly soluble in dichloromethane. Insoluble in water.
[B-](F)(F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2.[Rh]
373.991
373.99

Chemical Identifiers

36620-11-8
373.991
ZWPWNBFOFVFMPP-UHFFFAOYSA-N
10915722
[B-](F)(F)(F)F.C1C2C=CC1C=C2.C1C2C=CC1C=C2.[Rh]
C14H16BF4Rh-
MFCD00671775
bis bicyclo 2.2.1 hepta-2,5-diene rhodium tetrafluoroborate
bicyclo[2.2.1]hepta-2,5-diene;rhodium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

Documents