missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(neopentyl glycolato)diboron, 97%

$31.76 - $303.80

Chemical Identifiers

CAS 201733-56-4
Molecular Formula C10H20B2O4
Molecular Weight (g/mol) 225.886
MDL Number MFCD02093062
InChI Key MDNDJMCSXOXBFZ-UHFFFAOYSA-N
Synonym bis neopentyl glycolato diboron, bis neopentylglycolato diboron, 5,5,5',5'-tetramethyl-2,2'-bi 1,3,2-dioxaborinane, 2-5,5-dimethyl-1,3,2-dioxaborinan-2-yl-5,5-dimethyl-1,3,2-dioxaborinane, bis neopentylglycolato diboron, 5,5,5',5'-tetramethyl-2,2'-bi-1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 5,5,5',5'-tetramethyl, bis neopentyl glycolato diborane, bis 2,2-dimethyl-1,3-propanediolato diboron, 5,5,5',5'-tetramethyl-2,2' bi 1,3,2 dioxaborinanyl
PubChem CID 2734316
IUPAC Name 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)B2OCC(CO2)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1867503 View Documents Alfa Aesar™
L1867503
1g Each for $76.65
 
AAL1867506 View Documents Alfa Aesar™
L1867506
5g Each for $303.80
 
AAL18675MD View Documents Alfa Aesar™
L18675MD
250mg Each for $31.76
 
Specifications

Chemical Identifiers

201733-56-4
225.886
MDNDJMCSXOXBFZ-UHFFFAOYSA-N
2734316
B1(OCC(CO1)(C)C)B2OCC(CO2)(C)C
C10H20B2O4
MFCD02093062
bis neopentyl glycolato diboron, bis neopentylglycolato diboron, 5,5,5',5'-tetramethyl-2,2'-bi 1,3,2-dioxaborinane, 2-5,5-dimethyl-1,3,2-dioxaborinan-2-yl-5,5-dimethyl-1,3,2-dioxaborinane, bis neopentylglycolato diboron, 5,5,5',5'-tetramethyl-2,2'-bi-1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 5,5,5',5'-tetramethyl, bis neopentyl glycolato diborane, bis 2,2-dimethyl-1,3-propanediolato diboron, 5,5,5',5'-tetramethyl-2,2' bi 1,3,2 dioxaborinanyl
2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane

Specifications

170°C to 178°C
201733-56-4
MFCD02093062
bis neopentyl glycolato diboron, bis neopentylglycolato diboron, 5,5,5',5'-tetramethyl-2,2'-bi 1,3,2-dioxaborinane, 2-5,5-dimethyl-1,3,2-dioxaborinan-2-yl-5,5-dimethyl-1,3,2-dioxaborinane, bis neopentylglycolato diboron, 5,5,5',5'-tetramethyl-2,2'-bi-1,3,2-dioxaborinane, 2,2'-bi-1,3,2-dioxaborinane, 5,5,5',5'-tetramethyl, bis neopentyl glycolato diborane, bis 2,2-dimethyl-1,3-propanediolato diboron, 5,5,5',5'-tetramethyl-2,2' bi 1,3,2 dioxaborinanyl
B1(OCC(CO1)(C)C)B2OCC(CO2)(C)C
225.886
225.89
Bis(neopentyl glycolato)diboron
1g
C10H20B2O4
8839038
MDNDJMCSXOXBFZ-UHFFFAOYSA-N
2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-5,5-dimethyl-1,3,2-dioxaborinane
2734316
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C