Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(n-propyltetramethylcyclopentadienyl)strontium 1,2-dimethoxyethane adduct

$559.34 - $559.34

Chemical Identifiers

Chemical Name or Material Bis (n-propyltetramethylcyclopentadienyl)strontium 1,2-dimethoxyethane adduct
Molecular Formula C28H48O2Sr
Formula Weight 504.30g/mol (414.18g/mol)
MDL Number MFCD00269844
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4382406

 
alfa aesar™
43824-06
5g Each for $559.34
Description & Specifications

Specifications

Packaging Ampule
Quantity 5g
Sensitivity Air & Moisture Sensitive
Chemical Name or Material Bis (n-propyltetramethylcyclopentadienyl)strontium 1,2-dimethoxyethane adduct
Physical Form Liquid
Molecular Formula C28H48O2Sr
Formula Weight 504.30g/mol (414.18g/mol)
MDL Number MFCD00269844
Storage Note 1
  • Ampouled under argon

  • Ampouled under argon

Safety and Handling

  • Ampouled under argon