Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(n-propyltetramethylcyclopentadienyl)barium

Chemical Identifiers

Chemical Name or Material Bis(n-propyltetramethylcyclopentadienyl)barium
CAS 210758-43-3
Formula Weight 463.9
MDL Number MFCD00151385
UN Number UN1564
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3857402

 
alfa aesar™
3857402
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3857404

 
alfa aesar™
3857404
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Viscous Liquid/Solid
Formula Weight 463.9
Packaging Ampule
Chemical Name or Material Bis(n-propyltetramethylcyclopentadienyl)barium
MDL Number MFCD00151385
CAS 210758-43-3
UN Number UN1564
Quantity 0.5g
Health Hazard 1 Danger
Sensitivity Air & Moisture Sensitive
Health Hazard 2 GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P304+P340-P311-P301+P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.